Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025382.6 | NP_079658.2 | 6 - 7 | 1385 | 73 | |
GenBank mRNA | AY364165.1 | - | 6 - 7 | 1170 | 73 | |
AY846873.1 | - | 6 - 7 | 1386 | 73 | ||
AY846874.1 | - | 6 - 7 | 1386 | 73 | ||
BC019186.1 | - | 6 - 7 | 1354 | 73 | ||
BC043306.1 | - | 3 - 4 | 817 | 73 | ||
BC054355.1 | - | 7 - 8 | 1298 | 73 | ||
BC054769.1 | - | 6 - 7 | 1278 | 6415086 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |