Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164745.1 | NP_001158217.1 | 4 - 5 | 1021 | 69 | |
NM_008974.4 | NP_033000.1 | 4 - 5 | 1052 | 69 | ||
GenBank mRNA | AF035644.1 | - | 3 - 4 | 503 | 69 | |
AK033092.1 | - | 3 - 4 | 757 | 69 | ||
AK045093.1 | - | 3 - 4 | 754 | 69 | ||
AK135662.1 | - | 3 - 4 | 258 | 69 | ||
BC086794.1 | - | 3 - 4 | 434 | 6811716 | 69 | |
BC087551.1 | - | 5 - 6 | 1080 | 6329297 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |