Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145743.2 | NP_665686.2 | 4 - 5 | 681 | 86 | |
GenBank mRNA | AF397909.1 | - | 3 - 4 | 384 | 86 | |
AF520417.1 | - | 4 - 5 | 530 | 86 | ||
AK044595.1 | - | 4 - 5 | 557 | 86 | ||
AK170996.1 | - | 4 - 5 | 532 | 86 | ||
BC023308.1 | - | 3 - 4 | 358 | 86 | ||
BC089595.1 | - | 4 - 5 | 531 | 30118411 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |