Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081054.2 | NP_001074523.1 | 5 - 6 | 643 | 109 | |
GenBank mRNA | AK012351.1 | - | 5 - 6 | 644 | 109 | |
AK038801.1 | - | 5 - 6 | 652 | 109 | ||
AK041347.1 | - | 5 - 6 | 648 | 109 | ||
AK170225.1 | - | 5 - 6 | 588 | 109 | ||
BC070459.1 | - | 5 - 6 | 634 | 30605375 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |