Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400029632.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029632.mm.1 |
NM_001163027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029632.mm.1 |
NM_001163027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029632.mm.1 |
NM_198959.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029632.mm.1 |
NM_198959.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029634.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029634.mm.1 |
NM_001163027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029634.mm.1 |
NM_001163027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029634.mm.1 |
NM_198959.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029634.mm.1 |
NM_198959.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029631.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029631.mm.1 |
NM_001163027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029631.mm.1 |
NM_001163027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029631.mm.1 |
NM_198959.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029631.mm.1 |
NM_198959.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029637.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029637.mm.1 |
NM_198959.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029637.mm.1 |
NM_198959.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029628.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029628.mm.1 |
NM_001163027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029628.mm.1 |
NM_001163027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029628.mm.1 |
NM_198959.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029628.mm.1 |
NM_198959.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029627.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029627.mm.1 |
NM_001163027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029627.mm.1 |
NM_001163027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029627.mm.1 |
NM_198959.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029627.mm.1 |
NM_198959.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029630.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029630.mm.1 |
NM_001163027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029630.mm.1 |
NM_001163027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029630.mm.1 |
NM_198959.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029630.mm.1 |
NM_198959.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029636.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029636.mm.1 |
NM_001163027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029636.mm.1 |
NM_001163027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029638.mm.1 |
TC0400003626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029638.mm.1 |
NM_198959.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029638.mm.1 |
NM_198959.2 |
Refseq |