Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_001252250.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_001252250.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_001252251.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_001252251.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_001252252.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_001252252.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_001252253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_001252253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_010898.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
NM_010898.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020603.mm.1 |
TC1100002121.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020600.mm.1 |
NM_001252253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020600.mm.1 |
NM_001252253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020600.mm.1 |
TC1100002121.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_001252250.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_001252250.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_001252251.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_001252251.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_001252252.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_001252252.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_001252253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_001252253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_010898.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
NM_010898.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020602.mm.1 |
TC1100002121.mm.1 |
MTA TC |