Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243043.1 | NP_001229972.1 | 12 - 13 | 1982 | 56 | |
NM_001243044.1 | NP_001229973.1 | 12 - 13 | 1785 | 56 | ||
NM_007454.3 | NP_031480.2 | 12 - 13 | 1785 | 56 | ||
GenBank mRNA | AK034140.1 | - | 12 - 13 | 1985 | 56 | |
AK155892.1 | - | 12 - 13 | 1705 | 56 | ||
AK159273.1 | - | 12 - 13 | 1676 | 56 | ||
AK160042.1 | - | 12 - 13 | 1676 | 56 | ||
AK165747.1 | - | 12 - 13 | 1702 | 56 | ||
AK170376.1 | - | 12 - 13 | 1696 | 56 | ||
AK177753.1 | - | 2 - 3 | 161 | 56 | ||
BC008513.1 | - | 12 - 13 | 1598 | 3601435 | 56 | |
Y07919.1 | - | 12 - 13 | 1640 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |