Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144941.3 | NP_659190.3 | 5 - 6 | 1008 | 69 | |
GenBank mRNA | BC023677.1 | - | 5 - 6 | 983 | 5354497 | 69 |
BM944941.1 | - | 3 - 4 | 299 | 69 | ||
CV563343.1 | - | 4 - 5 | 579 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |