Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600009096.mm.1 |
TC0600001180.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009096.mm.1 |
NM_001109749.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009096.mm.1 |
NM_001109749.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009096.mm.1 |
NM_173004.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009096.mm.1 |
NM_173004.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009097.mm.1 |
TC0600001180.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009097.mm.1 |
NM_001109749.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009097.mm.1 |
NM_001109749.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009097.mm.1 |
NM_173004.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009097.mm.1 |
NM_173004.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009102.mm.1 |
TC0600001180.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009102.mm.1 |
NM_001109749.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009102.mm.1 |
NM_001109749.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009102.mm.1 |
NM_001109751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009102.mm.1 |
NM_001109751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009102.mm.1 |
NM_173004.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009102.mm.1 |
NM_173004.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009092.mm.1 |
TC0600001180.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009092.mm.1 |
NM_001109749.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009092.mm.1 |
NM_001109749.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009092.mm.1 |
NM_173004.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009092.mm.1 |
NM_173004.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009094.mm.1 |
TC0600001180.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009094.mm.1 |
NM_001109749.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009094.mm.1 |
NM_001109749.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009094.mm.1 |
NM_173004.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009094.mm.1 |
NM_173004.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009090.mm.1 |
TC0600001180.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009090.mm.1 |
NM_001109749.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009090.mm.1 |
NM_001109749.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009090.mm.1 |
NM_173004.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009090.mm.1 |
NM_173004.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009091.mm.1 |
TC0600001180.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009091.mm.1 |
NM_001109749.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009091.mm.1 |
NM_001109749.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009091.mm.1 |
NM_173004.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009091.mm.1 |
NM_173004.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009095.mm.1 |
TC0600001180.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009095.mm.1 |
NM_001109749.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009095.mm.1 |
NM_001109749.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009095.mm.1 |
NM_173004.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009095.mm.1 |
NM_173004.3 |
Refseq |