Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600006343.mm.1 |
TC0600000838.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006343.mm.1 |
NM_001253681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006343.mm.1 |
NM_001253681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006343.mm.1 |
NM_001253686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006343.mm.1 |
NM_001253686.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006343.mm.1 |
NM_029673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006343.mm.1 |
NM_029673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
TC0600000838.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253686.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253688.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253688.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253689.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_001253689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_029673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006345.mm.1 |
NM_029673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
TC0600000838.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253686.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253688.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253688.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253689.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_001253689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_029673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006346.mm.1 |
NM_029673.3 |
Refseq |