Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243075.1 | NP_001230004.1 | 3 - 4 | 319 | 132 | |
GenBank mRNA | AK012710.1 | - | 3 - 4 | 265 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |