Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100000801.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000801.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000801.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000806.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000806.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000806.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000807.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000807.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000807.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000795.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000795.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000795.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000804.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000804.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000804.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000803.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000803.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000803.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000802.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000802.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000802.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000809.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000809.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000809.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000805.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000805.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000805.mm.1 |
TC1100000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000808.mm.1 |
NM_023816.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000808.mm.1 |
NM_023816.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000808.mm.1 |
TC1100000074.mm.1 |
MTA TC |