Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007879.1 | NP_031905.1 | 7 - 8 | 962 | 111 | |
GenBank mRNA | AK044194.1 | - | 7 - 8 | 972 | 111 | |
AK080814.1 | - | 7 - 8 | 960 | 111 | ||
AK144438.1 | - | 7 - 8 | 964 | 111 | ||
AK155061.1 | - | 7 - 8 | 977 | 111 | ||
AK212980.1 | - | 1 - 2 | 64 | 111 | ||
AK217786.1 | - | 1 - 2 | 64 | 111 | ||
AK218042.1 | - | 1 - 2 | 64 | 111 | ||
AK218648.1 | - | 1 - 2 | 64 | 111 | ||
BC018596.1 | - | 3 - 4 | 392 | 111 | ||
BC039649.1 | - | 7 - 8 | 956 | 5351885 | 111 | |
D10715.1 | - | 7 - 8 | 962 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |