Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172154.1 | NP_001165625.1 | 3 - 4 | 499 | 75 | |
NM_027109.2 | NP_081385.1 | 3 - 4 | 616 | 75 | ||
GenBank mRNA | AK009174.1 | - | 3 - 4 | 622 | 75 | |
AK150242.1 | - | 3 - 4 | 529 | 75 | ||
BC023246.1 | - | 3 - 4 | 499 | 4039470 | 75 | |
DQ116784.1 | - | 3 - 4 | 510 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |