Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028000.1 | - | 1 - 2 | 47 | 62 | |
AK052762.1 | - | 4 - 5 | 1350 | 62 | ||
AK154613.1 | - | 4 - 5 | 1496 | 62 | ||
AK170641.1 | - | 4 - 5 | 1482 | 62 | ||
AK180272.1 | - | 1 - 2 | 122 | 62 | ||
BC007169.1 | - | 1 - 2 | 375 | 62 | ||
BC022976.1 | - | 4 - 5 | 1428 | 5362925 | 62 | |
BC024940.1 | - | 4 - 5 | 1425 | 62 | ||
BC071258.1 | - | 4 - 5 | 1361 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |