Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172784.3 | NP_766372.1 | 3 - 4 | 1116 | 60 | |
GenBank mRNA | AK031994.1 | - | 3 - 4 | 1058 | 60 | |
AK036676.1 | - | 3 - 4 | 1099 | 60 | ||
AK048919.1 | - | 3 - 4 | 1132 | 60 | ||
AK050544.1 | - | 3 - 4 | 1086 | 60 | ||
BC059874.1 | - | 3 - 4 | 937 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |