Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pithd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025411.4 | NP_079687.3 | 2 - 3 | 349 | 59 | |
GenBank mRNA | AF400670.1 | - | 2 - 3 | 312 | 59 | |
AK004208.1 | - | 2 - 3 | 350 | 59 | ||
AK050261.1 | - | 2 - 3 | 322 | 59 | ||
BC052695.1 | - | 2 - 3 | 249 | 5715038 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |