Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277121.1 | NP_001264050.1 | 9 - 10 | 1319 | 125 | |
NM_001277122.1 | NP_001264051.1 | 8 - 9 | 1158 | 125 | ||
NM_001277123.1 | NP_001264052.1 | 8 - 9 | 1153 | 125 | ||
NM_028871.2 | NP_083147.1 | 9 - 10 | 1322 | 125 | ||
GenBank mRNA | AF441128.1 | - | 8 - 9 | 1162 | 125 | |
AK046495.1 | - | 2 - 3 | 187 | 125 | ||
AK133840.1 | - | 8 - 9 | 1079 | 125 | ||
AK144682.1 | - | 9 - 10 | 1272 | 125 | ||
AK144992.1 | - | 8 - 9 | 1076 | 125 | ||
AK145849.1 | - | 8 - 9 | 1097 | 125 | ||
AK147043.1 | - | 9 - 10 | 1289 | 125 | ||
AK152039.1 | - | 9 - 10 | 1294 | 125 | ||
BC004679.1 | - | 8 - 9 | 1070 | 125 | ||
BC026850.1 | - | 9 - 10 | 1253 | 125 | ||
BC038051.1 | - | 9 - 10 | 1299 | 3500728 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |