Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010511.2 | NP_034641.1 | 1 - 2 | 239 | 106 | |
GenBank mRNA | AF128216.1 | - | 1 - 2 | 190 | 106 | |
AF128217.1 | - | 1 - 2 | 190 | 106 | ||
AK087525.1 | - | 1 - 2 | 228 | 106 | ||
AK150503.1 | - | 1 - 2 | 188 | 106 | ||
AK151079.1 | - | 1 - 2 | 193 | 106 | ||
AK152085.1 | - | 1 - 2 | 188 | 106 | ||
AK152384.1 | - | 1 - 2 | 188 | 106 | ||
AK153084.1 | - | 1 - 2 | 188 | 106 | ||
AK153347.1 | - | 1 - 2 | 191 | 106 | ||
AK159359.1 | - | 1 - 2 | 193 | 106 | ||
AK159660.1 | - | 1 - 2 | 133 | 106 | ||
AK159839.1 | - | 1 - 2 | 133 | 106 | ||
AK169593.1 | - | 1 - 2 | 208 | 106 | ||
AY157836.1 | - | 1 - 2 | 190 | 106 | ||
AY157837.1 | - | 1 - 2 | 190 | 106 | ||
BC132599.1 | - | 1 - 2 | 140 | 40130876 | 106 | |
BC138617.1 | - | 1 - 2 | 140 | 8861637 | 106 | |
BY309097.1 | - | 1 - 2 | 240 | 106 | ||
J05265.1 | - | 1 - 2 | 216 | 106 | ||
M25764.1 | - | 1 - 2 | 141 | 106 | ||
M26711.1 | - | 1 - 2 | 216 | 106 | ||
M28233.1 | - | 1 - 2 | 212 | 106 | ||
M28995.1 | - | 1 - 2 | 117 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |