Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010217.2 | NP_034347.2 | 1 - 2 | 349 | 84 | |
GenBank mRNA | BC006783.1 | - | 1 - 2 | 3589136 | 84 | |
BQ917383.1 | - | 1 - 2 | 288 | 84 | ||
CJ159336.1 | - | 1 - 2 | 350 | 84 | ||
M70642.1 | - | 1 - 2 | 84 | |||
M80263.1 | - | 1 - 2 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |