Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007939.2 | NP_031965.2 | 3 - 4 | 894 | 163 | |
GenBank mRNA | AK220393.1 | - | 1 - 2 | 378 | 163 | |
U72207.1 | - | 3 - 4 | 903 | 163 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |