Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017465.2 | NP_059493.2 | 2 - 3 | 305 | 95 | |
GenBank mRNA | AF026072.1 | - | 2 - 3 | 272 | 95 | |
AF478566.1 | - | 1 - 2 | 428 | 95 | ||
BB867946.1 | - | 2 - 3 | 306 | 95 | ||
BC009811.1 | - | 2 - 3 | 347 | 4008307 | 95 | |
BC009813.1 | - | 2 - 3 | 235 | 4039050 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |