Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291903.1 | NP_001278832.1 | 6 - 7 | 2821 | 68 | |
NM_009623.2 | NP_033753.2 | 6 - 7 | 2821 | 68 | ||
GenBank mRNA | AK138244.1 | - | 6 - 7 | 2822 | 68 | |
AK163443.1 | - | 6 - 7 | 2797 | 68 | ||
BC059010.1 | - | 6 - 7 | 1918 | 68 | ||
BC094231.1 | - | 6 - 7 | 2259 | 68 | ||
U85021.1 | - | 6 - 7 | 2681 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |