Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042491.2 | NP_001035956.1 | 6 - 7 | 901 | 101 | |
NM_001289517.1 | NP_001276446.1 | 6 - 7 | 901 | 101 | ||
NM_001289518.1 | NP_001276447.1 | 6 - 7 | 901 | 101 | ||
NM_001289519.1 | NP_001276448.1 | 6 - 7 | 901 | 101 | ||
NM_001289520.1 | NP_001276449.1 | 6 - 7 | 901 | 101 | ||
NM_021505.3 | NP_067480.2 | 6 - 7 | 901 | 101 | ||
GenBank mRNA | AK003821.1 | - | 3 - 4 | 294 | 101 | |
AK031432.1 | - | 6 - 7 | 826 | 101 | ||
AK049719.1 | - | 6 - 7 | 855 | 101 | ||
AK075969.1 | - | 6 - 7 | 840 | 101 | ||
AK088327.1 | - | 6 - 7 | 864 | 101 | ||
AK147951.1 | - | 6 - 7 | 840 | 101 | ||
AK148679.1 | - | 6 - 7 | 856 | 101 | ||
AK151314.1 | - | 6 - 7 | 847 | 101 | ||
AK151905.1 | - | 6 - 7 | 847 | 101 | ||
AK152528.1 | - | 6 - 7 | 817 | 101 | ||
AK152981.1 | - | 6 - 7 | 847 | 101 | ||
AK153373.1 | - | 6 - 7 | 847 | 101 | ||
AK153422.1 | - | 6 - 7 | 847 | 101 | ||
AK159204.1 | - | 6 - 7 | 847 | 101 | ||
AK159713.1 | - | 6 - 7 | 840 | 101 | ||
AK159916.1 | - | 6 - 7 | 840 | 101 | ||
AK167561.1 | - | 6 - 7 | 840 | 101 | ||
AK168737.1 | - | 6 - 7 | 841 | 101 | ||
AK168814.1 | - | 6 - 7 | 840 | 101 | ||
AK169196.1 | - | 6 - 7 | 864 | 101 | ||
AK215226.1 | - | 1 - 2 | 95 | 101 | ||
BC010339.1 | - | 6 - 7 | 817 | 2648173 | 101 | |
BC046566.1 | - | 6 - 7 | 880 | 2650790 | 101 | |
BC046804.1 | - | 6 - 7 | 838 | 6404923 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |