Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011537.3 | NP_035667.1 | 2 - 3 | 802 | 94 | |
GenBank mRNA | AF140427.1 | - | 2 - 3 | 567 | 94 | |
BC090639.1 | - | 2 - 3 | 806 | 30548234 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |