Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172721.2 | NP_766309.2 | 1 - 2 | 391 | 59 | |
GenBank mRNA | AK031815.1 | - | 1 - 2 | 385 | 59 | |
AK034447.1 | - | 1 - 2 | 319 | 59 | ||
AK047695.1 | - | 1 - 2 | 359 | 59 | ||
AK047756.1 | - | 1 - 2 | 333 | 59 | ||
AK053794.1 | - | 1 - 2 | 392 | 59 | ||
AK134291.1 | - | 1 - 2 | 398 | 59 | ||
BC009095.1 | - | 1 - 2 | 344 | 3158218 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |