Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177876.4 | NP_808544.1 | 2 - 3 | 327 | 71 | |
GenBank mRNA | AK082658.1 | - | 2 - 3 | 288 | 71 | |
AK166359.1 | - | 2 - 3 | 308 | 71 | ||
AK172114.1 | - | 2 - 3 | 288 | 71 | ||
AK172337.1 | - | 2 - 3 | 288 | 71 | ||
BC026744.1 | - | 2 - 3 | 328 | 4163734 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |