Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcwpw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005426.2 | NP_001005426.2 | 6 - 7 | 769 | 103 | |
GenBank mRNA | BC071186.1 | - | 6 - 7 | 696 | 30107853 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |