Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110827.2 | NP_001104297.1 | 8 - 9 | 1380 | 62 | |
NM_001110828.2 | NP_001104298.1 | 7 - 8 | 922 | 62 | ||
NM_001110829.2 | NP_001104299.1 | 7 - 8 | 922 | 62 | ||
NM_001110830.2 | NP_001104300.1 | 7 - 8 | 922 | 62 | ||
NM_001286417.1 | NP_001273346.1 | 7 - 8 | 922 | 62 | ||
NM_001286418.1 | NP_001273347.1 | 4 - 5 | 628 | 62 | ||
NM_001286419.1 | NP_001273348.1 | 6 - 7 | 826 | 62 | ||
NM_053104.6 | NP_444334.3 | 8 - 9 | 1380 | 62 | ||
NM_175387.3 | NP_780596.1 | 8 - 9 | 1380 | 62 | ||
GenBank mRNA | AF229055.1 | - | 7 - 8 | 865 | 62 | |
AF387322.1 | - | 7 - 8 | 666 | 62 | ||
AK044929.1 | - | 7 - 8 | 897 | 62 | ||
AK077594.1 | - | 8 - 9 | 1256 | 62 | ||
AK139555.1 | - | 9 - 10 | 1350 | 62 | ||
AY659951.1 | - | 7 - 8 | 666 | 62 | ||
AY659952.1 | - | 7 - 8 | 666 | 62 | ||
AY659953.1 | - | 4 - 5 | 372 | 62 | ||
AY904025.1 | - | 9 - 10 | 816 | 62 | ||
BC002124.1 | - | 7 - 8 | 760 | 3494630 | 62 | |
BC027263.1 | - | 8 - 9 | 932 | 3661748 | 62 | |
DQ017388.1 | - | 7 - 8 | 666 | 62 | ||
DQ017389.1 | - | 6 - 7 | 570 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |