Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133768.4 | NP_598529.1 | 4 - 5 | 379 | 78 | |
GenBank mRNA | AK076453.1 | - | 4 - 5 | 394 | 78 | |
AK138423.1 | - | 4 - 5 | 376 | 78 | ||
AK140160.1 | - | 4 - 5 | 394 | 78 | ||
AK148764.1 | - | 4 - 5 | 363 | 78 | ||
AK168739.1 | - | 4 - 5 | 374 | 78 | ||
BC016670.1 | - | 4 - 5 | 379 | 2648753 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |