Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133768.4 | NP_598529.1 | 7 - 8 | 607 | 78 | |
GenBank mRNA | AK010922.1 | - | 7 - 8 | 620 | 78 | |
AK076453.1 | - | 7 - 8 | 622 | 78 | ||
AK138423.1 | - | 7 - 8 | 604 | 78 | ||
AK140160.1 | - | 7 - 8 | 622 | 78 | ||
AK148764.1 | - | 7 - 8 | 591 | 78 | ||
AK168739.1 | - | 7 - 8 | 602 | 78 | ||
BC016670.1 | - | 7 - 8 | 607 | 2648753 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |