Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164311.1 | NP_001157783.1 | 12 - 13 | 2063 | 76 | |
NM_053083.3 | NP_444313.3 | 12 - 13 | 2060 | 76 | ||
GenBank mRNA | AF338440.1 | - | 11 - 12 | 1955 | 76 | |
AK014331.1 | - | 12 - 13 | 75 | |||
BC068307.1 | - | 12 - 13 | 2002 | 30059745 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |