Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026109.2 | NP_080385.2 | 1 - 2 | 178 | 94 | |
GenBank mRNA | AK019032.1 | - | 1 - 2 | 167 | 94 | |
BC132664.1 | - | 1 - 2 | 150 | 40130941 | 94 | |
BC132666.1 | - | 1 - 2 | 150 | 40130943 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |