Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044740.2 | NP_001038205.1 | 6 - 7 | 1267 | 67 | |
GenBank mRNA | AK154621.1 | - | 6 - 7 | 1241 | 67 | |
AK155558.1 | - | 6 - 7 | 1243 | 67 | ||
AK171369.1 | - | 6 - 7 | 1239 | 67 | ||
CX212155.1 | - | 2 - 3 | 351 | 67 | ||
M62838.1 | - | 6 - 7 | 1180 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |