Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF373710.1 | - | 4 - 5 | 218 | 79 | |
AK003572.2 | - | 4 - 5 | 248 | 79 | ||
AK003699.1 | - | 4 - 5 | 172 | 79 | ||
AK085669.1 | - | 4 - 5 | 297 | 79 | ||
BC013535.1 | - | 4 - 5 | 181 | 4239189 | 79 | |
BY083019.1 | - | 4 - 5 | 230 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |