Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164600.1 | NP_001158072.1 | 5 - 6 | 520 | 63 | |
NM_028364.2 | NP_082640.2 | 6 - 7 | 601 | 63 | ||
NM_133691.5 | NP_598452.2 | 5 - 6 | 550 | 63 | ||
GenBank mRNA | AK012434.1 | - | 5 - 6 | 490 | 63 | |
AK075960.1 | - | 5 - 6 | 518 | 63 | ||
AK149632.1 | - | 6 - 7 | 567 | 63 | ||
AK151526.1 | - | 5 - 6 | 514 | 63 | ||
AK168789.1 | - | 5 - 6 | 517 | 63 | ||
AK181470.1 | - | 1 - 2 | 126 | 63 | ||
BC010601.1 | - | 5 - 6 | 520 | 3257153 | 63 | |
BC119437.1 | - | 3 - 4 | 257 | 63 | ||
BC148274.1 | - | 3 - 4 | 257 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |