Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007964.2 | NP_031990.2 | 4 - 5 | 762 | 68 | |
GenBank mRNA | AK134294.1 | - | 4 - 5 | 963 | 68 | |
AK164899.1 | - | 1 - 2 | 87 | 68 | ||
AK165332.1 | - | 5 - 6 | 755 | 68 | ||
AK165885.1 | - | 4 - 5 | 715 | 68 | ||
AK192278.1 | - | 2 - 3 | 252 | 68 | ||
BC030056.1 | - | 5 - 6 | 769 | 68 | ||
BI737814.1 | - | 4 - 5 | 686 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |