Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145227.3 | NP_660262.2 | 6 - 7 | 1373 | 77 | |
GenBank mRNA | AB067535.1 | - | 6 - 7 | 1378 | 77 | |
AF418010.1 | - | 6 - 7 | 1313 | 77 | ||
BC118538.1 | - | 6 - 7 | 1313 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |