Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145226.2 | NP_660261.1 | 8 - 9 | 2022 | 65 | |
GenBank mRNA | AB067534.1 | - | 8 - 9 | 2022 | 65 | |
AF453830.1 | - | 8 - 9 | 2014 | 65 | ||
BC141055.1 | - | 8 - 9 | 2015 | 9055667 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |