Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145839.2 | NP_665838.1 | 6 - 7 | 951 | 69 | |
NM_181318.4 | NP_851835.1 | 6 - 7 | 948 | 69 | ||
GenBank mRNA | AK028756.1 | - | 6 - 7 | 840 | 69 | |
AK090085.1 | - | 6 - 7 | 843 | 69 | ||
AK149812.1 | - | 6 - 7 | 870 | 69 | ||
AY129963.1 | - | 6 - 7 | 878 | 69 | ||
BC050858.1 | - | 6 - 7 | 881 | 6517502 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |