Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080450.4 | NP_536698.2 | 1 - 2 | 98 | 61 | |
GenBank mRNA | AK034037.1 | - | 1 - 2 | 63 | 61 | |
AK045577.1 | - | 1 - 2 | 238 | 61 | ||
AK047368.1 | - | 1 - 2 | 99 | 61 | ||
AK147913.1 | - | 1 - 2 | 238 | 61 | ||
BC125305.1 | - | 1 - 2 | 85 | 40129820 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |