Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sart3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016926.1 | NP_058622.1 | 5 - 6 | 800 | 98 | |
GenBank mRNA | AF172722.1 | - | 5 - 6 | 800 | 98 | |
AK053828.1 | - | 5 - 6 | 804 | 98 | ||
AK086398.1 | - | 5 - 6 | 801 | 98 | ||
AK129067.1 | - | 5 - 6 | 795 | 98 | ||
BC036350.1 | - | 5 - 6 | 801 | 98 | ||
BC057156.1 | - | 5 - 6 | 791 | 5347469 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |