Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170851.1 | NP_001164322.1 | 3 - 4 | 364 | 148 | |
NM_008462.5 | NP_032488.4 | 3 - 4 | 364 | 148 | ||
GenBank mRNA | AF253057.1 | - | 2 - 3 | 198 | 148 | |
AF253058.1 | - | 2 - 3 | 198 | 148 | ||
AF253059.1 | - | 2 - 3 | 198 | 148 | ||
AF395446.1 | - | 2 - 3 | 198 | 148 | ||
AK017140.1 | - | 3 - 4 | 355 | 148 | ||
AY860971.1 | - | 2 - 3 | 198 | 148 | ||
BC064711.1 | - | 3 - 4 | 364 | 30295755 | 148 | |
EF025059.1 | - | 2 - 3 | 198 | 148 | ||
U10304.1 | - | 3 - 4 | 292 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |