Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145954.1 | NP_666066.1 | 2 - 3 | 250 | 65 | |
GenBank mRNA | AK030673.1 | - | 2 - 3 | 243 | 65 | |
AK144967.1 | - | 249 | 65 | |||
AK144995.1 | - | 2 - 3 | 257 | 65 | ||
AK151691.1 | - | 2 - 3 | 250 | 65 | ||
AK161198.1 | - | 2 - 3 | 288 | 65 | ||
AK164374.1 | - | 2 - 3 | 296 | 65 | ||
AK170276.1 | - | 2 - 3 | 289 | 65 | ||
AK220200.1 | - | 2 - 3 | 226 | 65 | ||
BC013548.1 | - | 2 - 3 | 250 | 4222058 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |