Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglecg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172900.3 | NP_766488.2 | 2 - 3 | 184 | 63 | |
GenBank mRNA | AK042488.1 | - | 2 - 3 | 185 | 63 | |
AK156766.1 | - | 2 - 3 | 226 | 63 | ||
AK162797.1 | - | 1 - 2 | 129 | 63 | ||
AY210400.1 | - | 1 - 2 | 39 | 63 | ||
BC114981.1 | - | 2 - 3 | 189 | 63 | ||
EF419770.1 | - | 1 - 2 | 47 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |