Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028236.1 | NP_082512.1 | 1 - 2 | 80 | 104 | |
GenBank mRNA | AK008389.1 | - | 1 - 2 | 112 | 104 | |
BC042780.1 | - | 1 - 2 | 80 | 4224563 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |