Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253916.1 | NP_001240845.1 | 10 - 11 | 2364 | 64 | |
NM_001253917.1 | NP_001240846.1 | 10 - 11 | 2364 | 64 | ||
NM_019982.3 | NP_064366.2 | 10 - 11 | 2364 | 64 | ||
GenBank mRNA | AB206790.1 | - | 10 - 11 | 2343 | 64 | |
AK083229.1 | - | 10 - 11 | 2328 | 64 | ||
AK122401.1 | - | 9 - 10 | 1478 | 64 | ||
BC056471.1 | - | 5 - 6 | 773 | 64 | ||
BC065117.1 | - | 10 - 11 | 2324 | 6840338 | 64 | |
BC065129.1 | - | 10 - 11 | 2141 | 30362651 | 64 | |
BC079593.1 | - | 4 - 5 | 698 | 64 | ||
BC094376.1 | - | 4 - 5 | 677 | 64 | ||
BC099839.1 | - | 3 - 4 | 485 | 64 | ||
DQ167195.1 | - | 10 - 11 | 2351 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |