Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133766.3 | NP_598527.2 | 21 - 22 | 2500 | 82 | |
GenBank mRNA | AB158474.1 | - | 21 - 22 | 2504 | 82 | |
AK030074.1 | - | 21 - 22 | 2501 | 82 | ||
AK032461.1 | - | 21 - 22 | 2473 | 82 | ||
AK044000.1 | - | 21 - 22 | 2494 | 82 | ||
AK044475.1 | - | 21 - 22 | 2496 | 82 | ||
AK129065.1 | - | 21 - 22 | 2478 | 82 | ||
BC007482.1 | - | 21 - 22 | 2504 | 3495608 | 82 | |
BC032092.1 | - | 21 - 22 | 2505 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |