Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK154877.1 | - | 1 - 2 | 190 | 69 | |
AK155880.1 | - | 1 - 2 | 171 | 69 | ||
AK166382.1 | - | 183 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |