Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177755.3 | NP_808423.1 | 2 - 3 | 1561 | 79 | |
GenBank mRNA | AK033353.1 | - | 2 - 3 | 1562 | 79 | |
BC132483.1 | - | 2 - 3 | 1463 | 40130760 | 79 | |
BC145690.1 | - | 2 - 3 | 1463 | 40130993 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |